ContactUsLinkComponent
Involuntary (Smooth) Muscle, Mammalian Slide
Involuntary (Smooth) Muscle, Mammalian Slide
# de catalogue 470177-590
Fournisseur:  Avantor
INVOLUNTARY (SMOOTH) MUSCLE (MAMMALIAN)
Involuntary (Smooth) Muscle, Mammalian Slide
# de catalogue 470177-590
Fournisseur:  Avantor
Spécifications
  • Description:
    Involuntary (Smooth) Muscle, Mammalian Slide
  • Cat. no.:
    470177-590
  • Stain:
    Hematoxylin and Eosin
  • Supplier No.:
    WARD470177-590
  • Orientation:
    Cross section & longitudinal section
Spécifications
A propos de cet article
Investigate smooth muscle characteristics
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Mammal. Entire small muscle.
Customers Who Bought This Also Bought