Corning® Microplates, 384-well, High Content Imaging Film Bottom, Corning
Supplier: Corning
Certificates
About this item
With an ultraclear film bottom and excellent flatness (whole plate and intra-well), these microplates are ideal for high resolution cellular imaging applications
- COC material allows for broad chemical resistance (including DMSO) and high mechanical stability
- Ultra-clear film with 127 µm thickness is well suited for imaging micropscopy
- Inter- and intra-well film bottom flatness within 50 and 10 µm, respectively, optimized for high content applications
- Low auto-fluorescence and birefringence
The microplate and film are manufactured from cyclic olefin copolymer, which has excellent optical properties, chemical resistance, and mechanical stability.
Specifications
- Catalog No:
- 75796-910
- 10037-554
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk