Corning® 96-Well High Content Screening Microplates with Film Bottom, Corning
Supplier: Corning
Certificates
About this item
With an ultraclear film bottom and excellent flatness (whole plate and intra-well), these microplates are ideal for high resolution cellular imaging applications.
- TC-treated, flat well bottom black microplate
- Made of Cyclic Olefin copolymer
- Ultraclear 190 µm film bottom thickness for imaging microscopy applications
- Low background fluorescence and minimal crosstalk allows highest possible optical quality for cell based assays
- Half area 96-well microplate reduces reagent consumption
- Sterile
- Inter and inter-well film bottom flatness optimized for high content applications
The microplate and film are manufactured from cyclic olefin copolymer, which has excellent optical properties, chemical resistance, and mechanical stability.
Specifications
- Catalog No:
- 75798-816
- 10037-552
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk