Spécifications
- Description:Salinity Test Kit with 40 Tests
- Cat. no.:470220-218
- Includes:Instructions
- Supplier No.:IS9755
Spécifications
A propos de cet article
How much Salt is Dissolved in a Body of Water?
- Easy activity to perform
- Gives clear results
This kit has been designed to allow the analyst to perform a very simple titration to determine the salinity of their water sample. The titration uses an easily identifiable color transition to indicate the end of the analysis.
Materials Included:
Silver Nitrate Titrant
Potassium Chromate
Plastic Pipettes
Reaction Vessels
Collection Bottles
Titrators
Measuring Tube