Spécifications
- Description:Green Fuel Cell Lab Activity
- No. of Lab Groups:15
- Cat. no.:470221-558
- Supplier No.:IS6500
Spécifications
A propos de cet article
The Green Fuel Cell Lab Activity is a great introduction to green energy production.
- Users Monitor the Voltage Generated by a Biological Fuel Cell
- Ideal Introduction to Green Energy Production
- Methylene Blue Works as an Electron Mediator
Create a biological fuel cell using the yeast Saccharomyces cerevisiae. A simple sugar is used as a food source for the yeast, and methylene blue is employed as an electron mediator. The cell is constructed without the mediator, and a baseline voltage is established as some electrochemical activity occurs.
Once the cell has stabilized, the mediator is added, and the effect on voltage is monitored and recorded using a voltage sensor. A voltage sensor is required.