Laboratory Jacks, Stainless Steel
Fournisseur: Wards
Certificats
A propos de cet article
Height-adjustable, corrosion-resistant stainless steel lab jacks capable of supporting heavy loads.
- Adjustable lab jacks with screw-operated mechanism for easy, fluid movement of platform
- Made of corrosion-resistant, easy-to-clean stainless steel
- Gas springs to support load
- Improved lift mechanism provides increased stability and torsional rigidity under load
- 10 mm tapped hole for accepting support rod
- Slip-proff rubber feet
- Can be autoclaved
Strong, stable support for laboratory apparatus. The lab jack has a screw-operated scissor action mechanism for easy, fluid movement of platform - with and without load - and precise height adjustments. For physical, chemical, biological experiments and other uses.
Customers Who Bought This Also Bought