To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Tubulin beta
- Clonality:Polyclonal
- Gene ID:203068
- Host:Rabbit
- Isotype:IgG
- Reactivity:Zebrafish,Hamster,Pig,Chicken,Human,Xenopus laevis,Rat,Mouse
- Antigen symbol:TUBB
- Conjugation:DyLight® 649
- Size:50 μl
- Antigen synonyms:tubulin beta polypeptide|tubulin beta-5 chain|M40|beta Ib tubulin|tubulin|beta 5-tubulin|tubulin beta chain|tubulin beta-1 chain|CDCBM6|beta polypeptide|beta-4 tubulin|OK/SW-cl.56|TUBB1|TUBB5
- Storage buffer:50mM sodium borate
- Storage temperature:Store at 4C. Do not freeze.
- Concentration:12 mg/ml
- Immunogen:A synthetic peptide made to the N-terminal region of human beta tubulin (within residues 1-100). Swiss-Prot P07437.
- Purification:IgG purified
- Cat. no.:10167-868
- Supplier No.:GTX48619
Spécifications
A propos de cet article
Rabbit Polyclonal antibody to beta Tubulin (DyLight 649)
Type: Primary
Antigen: TUBB
Clonality: Polyclonal
Clone:
Conjugation: DyLight649
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Chicken, Pig, Rat, Zebra Fish, Hamster, Xenopus laevis