ContactUsLinkComponent
The SpectraMax® Glo Steady-Luc™ Reporter Assay Kit, Molecular Devices
The SpectraMax® Glo Steady-Luc™ Reporter Assay Kit, Molecular Devices
# de catalogue 75801-460
Fournisseur:  Molecular Devices
Numéro CAS:  
SpectraMax* Glo Steady-Luc* Reporter Assay Kit
The SpectraMax® Glo Steady-Luc™ Reporter Assay Kit, Molecular Devices
# de catalogue 75801-460
Fournisseur:  Molecular Devices
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Format:
    Sufficient for (2) 96-well plates
  • Description:
    SpectraMax® Glo Steady-Luc™ Reporter Assay Explorer Kit
  • Cat. no.:
    75801-460
  • Supplier No.:
    R8352
Spécifications
A propos de cet article
The SpectraMax® Glo Steady-Luc™ Reporter Assay Kit provides a highly sensitive assay for the quantitation of firefly luciferase expression in mammalian cells.
  • Glow-type luminescence
  • Optimized for luminescence microplate readers
  • Simplified data acquisition and analysis
The SpectraMax® Glo Steady-Luc™ Reporter Assay Kit provides a highly sensitive assay for the quantitation of firefly luciferase expression in mammalian cells
Reporter gene assays are an important tool for biomedical and pharmaceutical researchers to monitor cellular events associated with gene expression, gene regulation, and signal transduction. Firefly luciferase, one of the most commonly used reporters, produces light by catalyzing a bioluminescence reaction that oxidizes D-luciferin in the presence of oxygen and ATP.
The application of luciferase-based reporter assay using luminescence microplate readers has become increasing popular for high-throughput analysis of gene expression as well as drug discovery applications. The SpectraMax Glo Steady-Luc Reporter Assay Kit is designed for optimal performance on microplate readers with high throughput enabled by both the microplate format and assay signal stability.
The assay kit is optimized for Molecular Devices SpectraMax® microplate readers with a preconfigured protocol provided in SoftMax® Pro Software for simplified data acquisition and analysis.
Customers Who Bought This Also Bought