Disposable Samplers, TargetDispo, Burkle
Fournisseur: BURKLE INC
Certificats
A propos de cet article
TargetDispo is used to take a target point sample at a position in the sample material that has previously been precisely determined.
- HDPE
- Clean room manufactured
- Optionally sterilised by gamma rays
The sampler pierces into the powder or granulate while closed. When it reaches the sample removal point it is opened and then closed again. In this way a precise target-point sample is obtained. The sample can simply be emptied through the open handle.
Customers Who Bought This Also Bought