Disposable Samplers, MicroDispo, Burkle
Fournisseur: BURKLE INC
Certificats
A propos de cet article
Using the MicroDispo disposable sampler, precise target point samples can be taken of free-flowing powders and small-grain bulk materials.
- Ergonomic handle
- Clean room manufactured
- Optionally sterilised by gamma rays
Quite small sample quantities can be taken with MicroDispo. Especially in pharmaceutical applications it often happens that only a small quantity can or should be extracted for analysis.
Customers Who Bought This Also Bought