ContactUsLinkComponent
ß-Hydroxybutyrate LiquiColor® Test, Stanbio Laboratory
ß-Hydroxybutyrate LiquiColor® Test, Stanbio Laboratory
# de catalogue 10152-474
Fournisseur:  Stanbio Laboratory
Numéro CAS:  
ß-Hydroxybutyrate LiquiColor® Test, Stanbio Laboratory
# de catalogue 10152-474
Fournisseur:  Stanbio Laboratory
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    ß-Hydroxybutyrate LiquiColor® Test (Endpoint)
  • Cat. no.:
    10152-474
  • Contents:
    Reagent 1: 1 x 50 mL; Reagent 2: 1 x 8.5 mL; Standard (1 mmol/L): 1 x 3 mL
  • Supplier No.:
    2440-058
Spécifications
A propos de cet article
Quantitative determination of ß-Hydroxybutyrate in human serum or plasma.
  • Available in two kit configurations
  • Determine presence and degree of ketosis
  • Linearity 4.5 mmol/L (46.8 mg/dL)
For the Quantitative determination of ß-Hydroxybutyrate in human serum or plasma. The presence and degree of ketosis can be determined by measuring blood levels of ß-hydroxybutyrate. This reagent is available in two kit configurations: a dual reagent kit for open channel chemistry analyzers and a 2 x 90 test kit that may be used directly on the Beckman Synchron® CX, LX and DX analyzers. Both kits include a 3 mL standard. Instrument specific applications are available from the manufacturer for open-channel automated and semi-automated chemistry analyzers. The ß-Hydroxybutyrate Reagent can also be used in a five minute procedure utilizing a spectrophotometer which reads absorbance at 505 nm wavelength.
Certifications: FDA 510(k). CE marked.
Ordering information: ß-Hydroxybutyrate tri-level control set, bi-level control set, and a linearity standard set are sold separately. A ß-Hydroxybutyrate validation pack and a Method Comparison kit are offered as resources to help laboratories get started using the ß-Hydroxybutyrate LiquiColor® test.
Caution: Store at 2 - 8º C until the expiration date on respective labeling.
Customers Who Bought This Also Bought