About this item
MPure FFPE DNA Extraction Kit is used with the MPure-12 Nucleic Acid Purification aNAP System for the extraction and purification of genomic DNA from FFPE (Formalin-Fixed, Paraffin-Embedded) tissue samples
- Extraction of genomic DNA from Formalin-Fixed, Paraffin-Embedded (FFPE) tissue samples
- Completely automated purification protocol
- Use with MPure-12 System
- Run in as little as 35 minutes
- Nucleic Acids can be used immediately in downstream applications
Nucleic acids extracted and purified from MPure FFPE DNA Extraction Kit can be used in a number of downstream application including: PCR; qPCR; Sequencing (NGS); Microarray; RFLP and; Southern Blot Analysis.
Capacity: 100-400 µL/ One to eight 10 µm- thick sections of tissue. 100-400 µL/ three to ten biopsies.