About this item
MPure Bacterial DNA Extraction Kit is used with the MPure-12 Nucleic Acid Purification System for extraction and purification of genomic DNA from both Gram-positive and Gram-negative bacteria.
- Extraction of genomic DNA from Gram-positive and Gram-negative bacteria
- Starting materials include bacterial pellet/colony from culture, cell-free body fluids, liquid transport media, urine, and environmental material
- Completely automated purification protocol
- Use with MPure-12 System
- Run in as little as 55 minutes
- Nucleic Acids can be used immediately in downstream applications
MPure™ Bacterial DNA Extraction Kit contains everything needed for sample preparation. However, for tough samples such as Gram+ bacteria or spores, soil or feces, sample preparation using FastPrep™ Instruments and Lysing Matrix Tubes can greatly increase yields as well as shorten processing time.
Capacity
Bacterial Colony: 200-400 µL /1-3 colony, Bacterial Pellet: 200-400 µL /Up to 109 bacteria (about OD600 = 3), cell-Free body fluids: 200-400 µL cell-free body fluids, Liquid Transport media: 200-400µL liquid transport media, Tissue: 200-400 µL /1-30 mg, Urine: 200-400 µL /5-50 mL urine