Viral/Pathogen Nucleic Acid Extraction Kit A, MP Biomedicals
Supplier: MP Biomedicals
Certificates
About this item
MPurify your Nucleic Acid: Obtain Superior Yield, Exceptional Purity, and Automated Performance in a Flexible, Convenient and Affordable Manner!
- Extract viral and bacterial DNA/RNA from cell-free samples
- Serum, plasma and other cell-free body fluids
- Use with MPure-12 System
- Run in as little as 40 minutes
- Nucleic Acid is ready for immediate use
The kit contains everything needed for sample preparation.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.