About this item
MPure Forensic DNA Extraction Kit is used with the MPure-12 Nucleic Acid Purification System for the extraction and purification of genomic DNA from forensic samples.
- Extract and isolate genomic DNA from forensic samples
- Completely automated purification protocol
- Use with MPure-12 System
- Run in as little as 40 minutes
- DNA is ready for immediate use
The kit contains everything needed for sample preparation. However, for tough samples such as swipes and swabs, cigarette butts and bodily fluid stained fabrics or other solid matrices, sample preparation using FastPrep™ Instruments and Lysing Matrix Tubes can greatly increase yields as well as shorten processing time. DNA extracted using the MPure Forensic DNA Extraction Kit is ready for immediate use and is compatible for quantitation using the Quantifiler™ Human, Quantifiler™ Y Human Male, Quantifiler™ Duo DNA Quantification Kits and Investigator™Quantiplex Kit, and for use in STR amplification using the AmpFISTR™ PCR Amplification kits as well as other downsteam applications.
Capacity. Chewing Gum - 40 mg, Cigarette butts - 1 cm2 paper from the end of the cigarette or filter, Clotted/ dried blood - 20 µL, Forensic Surface and Contact Swabs, Hair root - 0.5–1 cm, Human tissues - 40mg, Saliva - 50µL, Sperm Stains - 5-10 µL or 1 cm2 of the forensic sample