VWR® Thermal Printer Paper
Catalog # 47729-740
Supplier: VWR International
Specifications
- Description:Thermal printer paper
- Length:18 mm
- Cat. no.:47729-740
- Width:58 mm
- Packaging:1 Roll
Specifications
About this item
Heat sensitive paper for use with VWR® sterilizer with printer model AS12P.
Customers Who Bought This Also Bought