Life Latch® Screw Top UN Rated Poly Pail, PIG®, New Pig
Catalog # 10860-110
Supplier: New Pig
Specifications
- Description:Life Latch® Screw Top UN Rated Poly Pail
- Size:5 gallons
- Cat. no.:10860-110
- Supplier No.:DRM1054
Specifications
About this item
Open-head container is UN Rated for storing or transporting solid materials.
- Built-in locking mechanism
- Close lid simply and securely
- Chemical resistant
Built-in locking mechanism lets you close lid simply and securely. Open-head container is UN Rated for storing or transporting solid materials; can also be used for storing liquids. Durable HDPE construction is chemical resistant and FDA Approved.
Customers Who Bought This Also Bought