Poly Spill Deck, PIG®, New Pig
# de catalogue 10860-078
Fournisseur: New Pig
Spécifications
- Description:PIG® Poly Spill Deck
- Cat. no.:10860-078
- Sump Capacity:66 gal.
- Dimensions:50.06" W x 74.25" L x 5.19" H
- Supplier No.:PAK756-BK
Spécifications
A propos de cet article
The strongest standard drip deck you can buy for everyday drum storage.
- Wide, one-piece design
- Long-term storage of fully loaded drums
- Strongest standard drip deck
Wide, one-piece design eliminates the expense and labor of connecting multiple decks. Strong enough for long-term storage of fully loaded drums.
Customers Who Bought This Also Bought