Wobble-not Low-Insertion Force Serological Pipettes, VistaLab
Fournisseur: VistaLab
Certificats
A propos de cet article
These pipettes feature a unique 2-tier step design at the plug end for less jamming and less dripping.
- Sterile, RNase/DNase-free, Non-pyrogenic
- Individually Wrapped in Paper/Plastic or Plastic/Plastic
- Bulk packaging in plastic resealable bags
- Filter at plug end made of porous polypropylene
- Ultra-clear markings
Low-insertion force serological pipettes have an accuracy of ± 2% at full volume.
Customers Who Bought This Also Bought