PIG® Utility Containment Tray, New Pig
# de catalogue 10860-072
Fournisseur: New Pig
Spécifications
- Dimensions:33.75" W x 52" L x 5" H
- Description:PIG® Utility Containment Tray
- Cat. no.:10860-072
- Sump Capacity:29.92 gal.
- Supplier No.:PAK920
Spécifications
A propos de cet article
Durable, one-piece utility trays contain liquid messes from oily parts and leaky containers.
- Thick-wall, chemical-resistant polyethylene construction
- Ribs elevate parts to keep them away from trapped liquids
- Stackable trays store easily when not in use
Thick-wall, chemical-resistant polyethylene construction won't bend or warp. Ribs on bottom of tray add strength and elevate parts to keep them away from trapped liquids. Ideal for temporary storage of small containers that may leak or drip liquids onto the floor. Stackable trays store easily when not in use.
Customers Who Bought This Also Bought