Regulators for Scott/Air Liquide Transportable gases, Restek
Fournisseur: Restek
Certificats
A propos de cet article
Use the CGA-160 inlet connection with 14 L Scott/Air Liquide transportable gases. Use the CGA-165 inlet connection with 48 L Scott/Air Liquide transportable gases.
Customers Who Bought This Also Bought