Vacuum Gauges, Restek
Fournisseur: Restek
Certificats
A propos de cet article
High-quality vacuum gauges with 316 stainless steel wetted surfaces
- High quality
- For use with passive sampling kits
Recommended for use with passive sampling kits. All are rear mount.
–30" Hg to 0" Hg.
Customers Who Bought This Also Bought