Humidification Chamber, Restek
# de catalogue 10862-928
Fournisseur: Restek
Spécifications
- Description:Humidification Chamber
- Cat. no.:10862-928
- Supplier No.:24282
Spécifications
A propos de cet article
Restek’s humidification chamber is made of acrylic and withstands pressure up to 90 psi.
- Withstands high pressures
- Acrylic
- Easy connection to pressure lines
When cleaning SilcoCan® or TO-Can® canisters, it is important to use humidified air or nitrogen to help remove volatile organic contaminants. Restek’s humidification chamber is made of acrylic and withstands pressure up to 90 psi. The 1/4-inch inlet and outlet compression fittings allow easy connection to pressure lines on your cleaning system. Our humidification chamber also has an easy-to-open lid for filling with water.
Customers Who Bought This Also Bought