PUF Glass Holders, Restek
Fournisseur: Avantor
Certificats
A propos de cet article
Durable and reusable, PUF glass holders reduce waste and a cost-effective alternative to precleaned packed cartridges.
- Fit either 6 cm or 22 mm O-Ø PUF plug or can be used with bulk SDVB resin
- Large glass holder fitted with double stainless steel screens for support
- Small glass holder has stem designed for secure 1/4" I-Ø tubing connection to sampling pump
Customers Who Bought This Also Bought