Cleaned Polyurethane Foam (PUF) Cartridges, Restek
Fournisseur: Restek
Certificats
A propos de cet article
These cartridges used for collection of semivolatiles (pesticides, PCBs, PAHs).
- Precleaned and ready to use
- Suitable for ambient, indoor, and industrial hygiene applications
- PUF/XAD-2 "sandwiches" capture a wider range of semivolatiles
Both large high-volume (220 to 280 l/min) and small low-volume (1 to 5 l/min) PUFs available.
Customers Who Bought This Also Bought