Canister Carrying Case, Restek
# de catalogue 10862-808
Fournisseur: Restek
Spécifications
- Description:Carrying Case Deluxe, for 2x6L canister
- Cat. no.:10862-808
- Supplier No.:24226
Spécifications
A propos de cet article
Fits two 6 L SilcoCan® or TO-Can® canisters tightly without foam.
- Heavy-duty
- All-aluminum design
No organic contaminants from foam or plastics.
Customers Who Bought This Also Bought