Aura™ Personal Air Samplers (Disposable 8-Hour Sampler Lines), Restek
Fournisseur: Restek
Certificats
Certificats
A propos de cet article
Customers Who Bought This Also Bought
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
$118.61 to $1,128.11
MicroSolv™ Pharma Grade Vials, Caps and Convenience Kits
Fournisseur: MicroSolv Technology Corp
$116.48
SureSTART™ Glass Crimp Top Vials, 2 ml, Level 2 High-Throughput Applications, Thermo Scientific
Fournisseur: Thermo Scientific
$1,347.98 to $1,347.98
WHEATON® MicroLiter Sample Vial Component Kits, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$467.43
SureSTART™ Crimp Caps, 20 mm, Level 2 High-throughput Applications, Thermo Scientific
Fournisseur: Thermo Scientific
$11,110.75 to $11,110.75
SureSTART™ Septa for 20 mm Stoppers, Level 2 High-throughput Applications, Thermo Scientific
Fournisseur: Thermo Scientific
$131.41 to $136.31
SureSTART™ Glass Inserts for 2 ml Vials, Level 3 High Performance Applications, Thermo Scientific
Fournisseur: Thermo Scientific
$120.60 to $125.07