Sample Inlets (Stainless Steel & Siltek® Treated), Restek
Fournisseur: Restek
Certificats
A propos de cet article
Sample inlets with fittings on each end.
- Inlets have 1/4" stainless steel compression fitting on each end
- One end connects to flow controller or canister
- Nut on other end serves as rain guard
- Includes nuts and ferrules
- Two different lengths for use with large canisters and miniature canisters
Customers Who Bought This Also Bought