Passive Air Sampling Kits-Soil Gas (Stainless Steel and Siltek®-Treated), Restek
Fournisseur: Restek
Certificats
A propos de cet article
This unique grab sampler is specifically designed for soil gas sampling by allowing the connection of tubing coming from the soil gas sample port.
- Stainless steel tee with orifice
- Variety of orifice sizes for sampling from 4 minutes to 10 hours
- 1 1/2" vacuum gauge (–30" Hg to 0" Hg)
The innovative design minimizes connections and leaks and houses a critical orifice in the tee fitting. It also incorporates a vacuum gauge and 2 µm frit filter. The 1/4" compression inlet and outlet allows easy connection to the canister valve and also to the tubing from the sample port. Several orifice sizes provide sampling times from 4 minutes to 10 hours on a 6 L canister. Individual replacement parts are available, providing a cost-effective alternative to replacing the entire sampler.
BodyTextAccessories: Air sampling canisters sold separately.
Spécifications
- Catalog No:
- 10862-718
- 10862-692
- 10862-690
- 10862-682
- 10863-168
- 10862-768
- 10863-196
- Flow rate:
- 80 mL/min
- 19 mL/min
- 6 mL/min
- 150 mL/min
- 210 mL/min
- 30 mL/min
- 150 mL/min