Miniature Air Sampling Kits (Stainless Steel and Siltek®-Treated), Restek
Fournisseur: Restek
Certificats
A propos de cet article
Restek’s passive air sampling kit incorporates all the hardware necessary to collect air samples and is easy to assemble for field sampling.
- Provide accurate integrated sampling without a sampling pump
- Convenient smaller size connects easily to miniature canisters
- Available in stainless steel or with Siltek® treatment for greater inertness
BodyTextAccessories: Air sampling canisters sold separately.
Ordering information: Kit includes flow controller, critical orifice, 2 µm frit filter, vacuum gauge, and sample inlet. The gauge (cat.# 24120) and sample inlet (cat.#s 26211, 26212) are downsized for use with smaller canisters.
Spécifications
- Catalog No:
- 10862-872
- 10862-874
- 10863-030
- 10862-970
- 10862-612
- 10862-974
- 10862-976
- Flow rate:
- 0.5–2 mL/min
- 4–8 mL/min
- 2–4 mL/min
- 0.5–2 mL/min
- 8–15 mL/min
- 4–8 mL/min
- 8–15 mL/min