Veriflo® Flow Controllers (Stainless Steel and Siltek® Treated), Restek
Fournisseur: Restek
Certificats
A propos de cet article
Veriflo® 423XL flow controllers are offered in a Siltek®-treated and stainless steel version.
- Treated and non-treated version
- Available with or without critical orifice
The flow device is available with or without a critical orifice. The critical orifice in a Veriflo® flow controller is interchangeable.
BodyTextAccessories: Order orifices for alternate sampling times, or replacement orifices, separately.
Ordering information: Vacuum gauge sold separately.
Customers Who Bought This Also Bought