Passive Air Sampling Kits-Integrated (Stainless Steel and Siltek®-Treated), Restek
Fournisseur: Restek
Certificats
A propos de cet article
Restek’s passive air sampling kit incorporates all the hardware necessary to collect air samples and is easy to assemble for field sampling.
- Provide accurate integrated sampling without a sampling pump
- Siltek®-treated components ensure accurate sampling of active components
- Excellent for sampling times from 0.5 hour to 125 hours
The improved filter design greatly reduces the number of potential leak sites.The passive air sampling kit is available in seven sampling flow ranges and in stainless steel or Siltek®-treated finish. The stainless steel kit is ideal to partner with the Restek® TO-Can® air sampling canister for TO-14A and TO-15 methods. Use the Siltek®-treated version with the Restek® SilcoCan® air sampling canister when collecting low-level volatile sulfur compounds or other active compounds.
BodyTextAccessories: Vacuum/pressure gauge included in kit; air sampling canisters sold separately.
Spécifications
- Catalog No:
- 10862-738
- 10863-098
- 10862-580
- 10862-754
- 10862-966
- 10862-616
- 10862-614
- 10863-060
- 10863-062
- 10863-064
- Flow rate:
- 8–15 mL/min
- 0.5–2 mL/min
- 80–340 mL/min
- 2–4 mL/min
- 80–340 mL/min
- 4–8 mL/min
- 2–4 mL/min
- 4–8 mL/min
- 15–30 mL/min
- 30–80 mL/min