Passive Air Sampling Kits-Grab (Stainless Steel and Siltek®-Treated), Restek
Fournisseur: Restek
Certificats
A propos de cet article
Passive air sampling kits for use with 1, 3, or 6 L canisters for grab air sampling.
- Use with 1, 3, or 6 L canisters for grab air sampling
- Variety of orifice sizes for fast sampling from 5 to 60 minutes
- Connect 1/4" compression fitting directly to canister valve inlet
- Replaceable frit protects orifice and valve from particulates
BodyTextAccessories: Air sampling canisters sold separately.
Spécifications
- Catalog No:
- 10862-600
- 10862-572
- 10862-594
- 10863-012
- 10862-570
- 10862-592
- 10863-014
- 10862-568
- 10862-590
- 10863-016
- 10862-588
- 10863-018
- 10863-150
- 10862-912
- 10862-604
- 10862-602
- Flow rate:
- 15 mL/min
- 80 mL/min
- 80 mL/min
- 150 mL/min
- 45 mL/min
- >1,000 mL/min
- 300 mL/min
- 15 mL/min
- 390 mL/min
- 390 mL/min
- 300 mL/min
- >1,000 mL/min
- 150 mL/min
- 20 mL/min
- 45 mL/min
- 20 mL/min