Steel Loading Ramp With Non-Slip Poly Grate, PIG®, New Pig
# de catalogue 10860-062
Fournisseur: New Pig
Spécifications
- Description:PIG® Steel Loading Ramp With Non-Slip Poly Grate
- Cat. no.:10860-062
- Dimensions:26" W x 67" L
- Supplier No.:PAK603
Spécifications
A propos de cet article
Combination of steel frame with polyethylene grate surface is stronger than all-poly, yet lighter than all-steel construction.
- Hinge assemblies
- Metal plate secures ramp to pallet or deck
- Nonslip poly grate surface provides traction for added safety
Hinge assemblies let you store ramp upright inside a PIG Roll Top Hardcover. Metal plate secures ramp to pallet or deck and provides a smooth transition for loading and unloading. Nonslip poly grate surface provides traction for added safety. Edge guards guide drum trucks and help prevent them from rolling off the sides. Note: Ramp must be detached to close doors.
Customers Who Bought This Also Bought