Microcentrifuge Tubes with Snap Cap, Bio Plas
Fournisseur: Bio Plas
Certificats
A propos de cet article
The Snap-Cap Microcentrifuge Tube gives a firm, dependable seal, and its tough virgin-polypropylene construction withstands the rugged G-forces of high-speed centrifugation.
- Ultra low retention
- Snap-Caps for optimal sealing
- RNase and DNase free and non-pyrogenic
Used for specimen collection, mixing, electrophoresis sampling/handling, enzyme assays, radioassays, immumoassays and many other applications. The Snap-Cap Microcentrifuge Tube’s smooth, conical interior lets remove entire sample with ease, and without material hangup. Fits Beckman™, Eppendorf™ and all other microcentrifuges.
The Snap-Cap is attached to each tube, and the entire unit is autoclavable at 250°F
at 15 psi for 15 minutes.
Customers Who Bought This Also Bought