Digital Stopwatch, Industrial Physics
# de catalogue 10025-184
Fournisseur: GREAT LAKES GAGES, LLC
Spécifications
- Description:Digital stopwatch
- Cat. no.:10025-184
- Dimensions:3.15×0.71×2.36"
- Weight:1.6 oz.
- Supplier No.:DI0076
Spécifications
A propos de cet article
Accurate digital TQC stopwatch, resolution to ¹/₁₀₀th of a second.
- Split times, built-in alarm
- Clear display with large digits
- Indispensible for viscosity measurements
Ultra flat and robust water- and shock proof design, complete with carrying cord.
Customers Who Bought This Also Bought