Drum Dolly, Stainless Steel, Wesco
Fournisseur: Wesco
Certificats
A propos de cet article
Made of Stainless Steel dolly construction with reinforced outriggers.
- Ideal for clean room, pharmaceutical, food and wash down applications
- Swivel rig with 3" hard rubber wheel and stainless hardware
Customers Who Bought This Also Bought