Vented Screw Caps, Non sterile, Chemglass
# de catalogue 30623-042
Fournisseur: Chemglass
Small Business Enterprise
Spécifications
- Sterility:Non sterile
- Description:Vented Screw Cap, Gray
- Supplier No.:CLS-1484-10
Spécifications
A propos de cet article
Polypropylene vented cap has a 0.22 µm (PTFE) hydrophobic membrane to allow gas exchange while preventing contamination.
- Autoclavable
- Liner-less plug seal design for efficient sealing
- GL-45 screw thread type
Customers Who Bought This Also Bought