Test Tube Brushes, Justman
Fournisseur: Justman Brush
Certificats
A propos de cet article
Test tube brush is especially designed to clean the inner walls and bottom of test tubes to remove every trace of the chemical stored in it.
- White-colored bristle with silver handle
- Test tube brush with a radial tip
The tip of this brush features formed sprays that thoroughly clean the bottom of the test tube without damaging or scratching the glassware.
Customers Who Bought This Also Bought