Reversible Compound Kater's Pendulum
# de catalogue 470220-608
Fournisseur: OSAW INDUSTRIAL PRODUCTS SE
Spécifications
- Description:Reversible Compound Kater's Pendulum
- Cat. no.:470220-608
- Supplier No.:SP272
Spécifications
A propos de cet article
Unlike traditional pendulums, the Kater's pendulum center of gravity does not have to be know in order to determine the acceleration due to gravity.
- Sturdy design
- Repeatable results
This pendulum consists of a electroplated brass rod (120 cm long, 10 mm diameter), one pair each of small and large adjustable brass and wooden masses, two steel knife edges set in adjustable metal holders and a metal support bracket for mounting on walls.
Customers Who Bought This Also Bought