VWR® Numbered Animal Ear Tags
# de catalogue 10806-296
Fournisseur: VWR International
Spécifications
- Description:Aluminum numbered Animal Ear Tags
- Cat. no.:10806-296
Spécifications
A propos de cet article
Ear tags are made from an ASTM standard (B209-10) aluminum sheet.
- Aluminum sheet construction
- Easy to insert
- Numbered for quick identification
The small aluminum tags are numbered 1 to 100, and are supplied with 100 washers. To apply, simply insert tag through hole and place washer behind hole. Bend tag ends back in rivet fashion, leaving space for ear growth.
Customers Who Bought This Also Bought