ContactUsLinkComponent
EXP® Hand-Tight Coupler (EXP® Reusable Fittings for HPLC and UHPLC for 10-32 fittings and ¹/₁₆" tubing), Restek
EXP® Hand-Tight Coupler (EXP® Reusable Fittings for HPLC and UHPLC for 10-32 fittings and ¹/₁₆" tubing), Restek
# de catalogue 10853-598
Fournisseur:  Restek
EXP* Hand-Tight Coupler
EXP® Hand-Tight Coupler (EXP® Reusable Fittings for HPLC and UHPLC for 10-32 fittings and ¹/₁₆" tubing), Restek
# de catalogue 10853-598
Fournisseur:  Restek
Spécifications
  • Description:
    EXP Hand-Tight Coupler, 2 Hand-Tight Nuts, 2 Hybrid Ferrules, ea.
  • Cat. no.:
    10853-598
  • Supplier No.:
    25940
Spécifications
A propos de cet article
Restek is pleased to offer the reusable EXP® fitting system from Optimize Technologies for the ultimate in easy, reliable LC connections.
  • Hand-tight fitting style achieves effortless HPLC seals—no tools needed for a 8,700+ psi seal
  • Both hand-tight and hex-head styles wrench-tighten for reliable UHPLC use up to 20,000+ psi
  • Patented ferrule can be installed repeatedly without compromising high-pressure seal
  • Hybrid design combines the durability of titanium with the sealing ability of PEEK
  • Cutting-edge system provides ZDV (zero dead volume) connection to any 10-32 female port
  • Compatible with 1/16" PEEK and stainless steel tubing
The patented hybrid EXP® ferrule combines the durability of titanium with the sealing ability of PEEK for a swage that can be reused over and over again. And, when you choose the hand-tight fitting style, the special EXP® nut offers an effortless seal up to 8,700+ psi (600+ bar)—no tools needed! For a reliable 20,000+ psi (1,400+ bar) UHPLC connection with either fitting style, simply wrench-tighten an extra 1/4 to 1/2 turn. EXP® ferrules should only be used with genuine EXP® nuts. When used with an EXP® nut, the EXP® ferrule provides repeated ZDV (zero dead volume) connections to any 10-32 female threaded port, including Restek® LC columns, 6-port injection valves, and more. To accommodate varying port depths, simply hold the tubing fully bottomed in the port and tighten as instructed.
Ordering information: Hybrid Ferrule U.S. Patent No. 8201854, Optimize Technologies. Optimize Technologies EXP Holders are Patent Pending. Other U.S. and Foreign Patents Pending. The Opti- prefix is a registered trademark of Optimize Technologies, Inc. Do not use EXP® ferrules with standard nuts. Failure to use EXP® fittings according to the included instructions may result in unsafe UHPLC connections and/or non-ZDV connections.
Customers Who Bought This Also Bought