Spécifications
- Description:EXP Hand-Tight Coupler, 2 Hand-Tight Nuts, 2 Hybrid Ferrules, ea.
- Cat. no.:10853-598
- Supplier No.:25940
Spécifications
A propos de cet article
Restek is pleased to offer the reusable EXP® fitting system from Optimize Technologies for the ultimate in easy, reliable LC connections.
- Hand-tight fitting style achieves effortless HPLC seals—no tools needed for a 8,700+ psi seal
- Both hand-tight and hex-head styles wrench-tighten for reliable UHPLC use up to 20,000+ psi
- Patented ferrule can be installed repeatedly without compromising high-pressure seal
- Hybrid design combines the durability of titanium with the sealing ability of PEEK
- Cutting-edge system provides ZDV (zero dead volume) connection to any 10-32 female port
- Compatible with 1/16" PEEK and stainless steel tubing
The patented hybrid EXP® ferrule combines the durability of titanium with the sealing ability of PEEK for a swage that can be reused over and over again. And, when you choose the hand-tight fitting style, the special EXP® nut offers an effortless seal up to 8,700+ psi (600+ bar)—no tools needed! For a reliable 20,000+ psi (1,400+ bar) UHPLC connection with either fitting style, simply wrench-tighten an extra 1/4 to 1/2 turn. EXP® ferrules should only be used with genuine EXP® nuts. When used with an EXP® nut, the EXP® ferrule provides repeated ZDV (zero dead volume) connections to any 10-32 female threaded port, including Restek® LC columns, 6-port injection valves, and more. To accommodate varying port depths, simply hold the tubing fully bottomed in the port and tighten as instructed.