Lamps for Agilent HPLC Systems, Restek
Fournisseur: Restek
Certificats
Certificats
A propos de cet article
Lamps are compatible with HPLC system.
Customers Who Bought This Also Bought
Glycerine ≥99.7%, Laboratory Reagent, VWR Chemicals BDH®
Fournisseur: VWR International
$881.80 to $2,807.56
Sodium sulfate, anhydrous ≥99.0%, white powder ACS, VWR Chemicals BDH®
Fournisseur: VWR International
$290.42 to $873.69
VeriSpec™ Potassium - K⁺ Standard for Ion Chromatography 1000 ppm in H₂O, RICCA Chemical Company
Fournisseur: Ricca Chemical
$170.97
1-Octanesulfonic acid sodium salt ≥99%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Fournisseur: VWR International
$814.96
Dichloromethane ≥99.9% (by GC), ChromAR® for HPLC, for UV spectrophotometry, Macron Fine Chemicals™
Fournisseur: AVANTOR PERFORMANCE MATERIAL LLC
$137.00 to $822.14
Sorensen's Phosphate Buffer, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
$86.00