Seal Pack (Rebuilt with Needle) for Waters 717 Autosampler, Restek
# de catalogue 10853-804
Fournisseur: Restek
Spécifications
- Description:Seal Pack (Rebuilt) with Needle
- Cat. no.:10853-804
- Supplier No.:25496
Spécifications
A propos de cet article
Delivery: Kit includes seal pack assembly, pre-column filter assembly, needle, 0.020" ID tube assembly, compression screw, ferrule.
Customers Who Bought This Also Bought