Seal Pack Rebuild Kit (without Seal Wash Tube Assembly) for Waters HPLC Systems, Restek
# de catalogue 10853-646
Fournisseur: Restek
Spécifications
- Description:Seal Pack Rebuild Kit, for Alliance 2690 HPLC System
- Cat. no.:10853-646
- Supplier No.:25495
Spécifications
A propos de cet article
Delivery: Kit includes wash tube seal, seal wash tube, PTFE washer, filter retainer, lower wash seal frit, needle wash frit, TFE washer, needle, injector seal (2), compression screw, ferrule.
Customers Who Bought This Also Bought