Inlet Manifold Kit for Waters HPLC Systems, Restek
# de catalogue 10853-710
Fournisseur: Avantor
Spécifications
- Description:Inlet Manifold Kit for Waters M45, 501,510, 590, 600(E), Similar to Waters #WAT060448
- Model:M45, 501, 510, 515, 590, 600, 610
- Includes:TFE ball plug, TFE seat, ruby ball, manifold insert, TFE insert seal, Belleville washer (2), flat washer, inlet tube body assembly
- Supplier No.:25412