Check Valve Cartridges for Waters HPLC Systems, Restek
# de catalogue 10853-584
Fournisseur: Restek
Spécifications
- Description:Check Valve Cartridges (2) for Waters Alliance, Similar to Waters #WAT270941, 2-pk.
- Model:2690, 2695, 2790, 2795
- Cat. no.:10853-584
- Supplier No.:25373