ContactUsLinkComponent
Deuterium Lamps for Shimadzu HPLC Systems, Restek
Deuterium Lamps for Shimadzu HPLC Systems, Restek
Catalog # 10853-462
Supplier:  Restek
Deuterium Lamp, for HPLC Systems, Model Number: SPD-10, 10A, 10AVvp, SPD-20A, SPD-20AV
Deuterium Lamps for Shimadzu HPLC Systems, Restek
Catalog # 10853-462
Supplier:  Restek
Specifications
  • Description:
    Deuterium Lamp for Shimadzu SPD-10A, 10AV, SPD-20A Similar to Shimadzu# 228-34016-02
  • Cat. no.:
    10853-462
  • Model:
    SPD-10, 10A, 10AVvp, SPD-20A, SPD-20AV
  • Supplier No.:
    25284