Outlet Check Valves for Shimadzu HPLC Systems, Restek
Fournisseur: Restek
Certificats
Certificats
A propos de cet article
Customers Who Bought This Also Bought
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
50,14 $ to 501,54 $
VWR® Screw Caps for 9 mm Screw-Thread Vials, Wide Opening
Fournisseur: VWR International
57,65 $ to 603,92 $
WHEATON® MicroLiter Crimp-Top Headspace Vials, 20 mm, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
109,95 $
Septa Only, GPI 20-400, Gray PTFE/Silicone, Dimensions: 20mm x 0.060 inch, for Screw Thread Closure
Fournisseur: Chemglass
113,00 $
VWR® 11 mm Crimp Top Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
291,56 $
SureSTART™ GOLD-Grade Glass Insert for 2 ml Vials, Level 3 High Performance Applications, Thermo Scientific
Fournisseur: Thermo Scientific
810,00 $ to 840,15 $
AQR™ Caps for RSA™ Autosampler Vials, MicroSolv Technology Corporation
Fournisseur: MicroSolv Technology Corp
676,79 $
WHEATON® MicroLiter µLPlate® Component Kits, Inserts and Caps, Unassembled, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
184,08 $
WHEATON® MicroLiter 20 mm Crimp Seals with Septa, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
211,44 $