Ferrules for Secure-Fit, ¹/₁₆", Restek
Fournisseur: Restek
Certificats
Customers Who Bought This Also Bought
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
$118.61 to $1,128.11
VWR® Screw Caps for 9 mm Screw-Thread Vials, Wide Opening
Fournisseur: VWR International
$79.43 to $662.05
BASIK* 1.5 ml Snaptop vial and cap slit eppendorf amber
Fournisseur: MicroSolv Technology Corp
$115.66
VWR® 11 mm Crimp Top Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
$279.61
WHEATON® MicroLiter 11 mm Crimp-Top Seals, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$79.24